Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Bromotoluene 99%

2-Bromotoluene 99%
 
2-Bromotoluene 99%
2-Bromotoluene 99%

 

 
  •  :95-46-5
  •  :C₇H₇Br
  •  :MFCD00000068
  •  :171.04 g/mol
  •  :13,01424
  •  :181.5 °C (1013 hPa)
  •  :–27 °C
  •  :1.42 g/cm³
  •  :78 °C
  •  :Refrigerator
  •  :202-421-7
  •  :9,III

 

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5550 to 1.5570 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    25 mL
       
       
    1 * 25 mL
    1 * 25 mL - 32,10 €
    32,10 €
    1 * 25 mL
       32,10 €

  • Glass bottle
    100 mL
       
       
    1 * 100 mL
    1 * 100 mL - 87,30 €
    87,30 €
    1 * 100 mL
       87,30 €