11-Chloro-1-undecene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:872-17-3
:
Formula:C₁₁H₂₁Cl
MDL Number:MFCD00671354
Boiling Pt:108…109 °C (6 mmHg)
Density:0.995
Flash Pt:104 °C (219 °F)
:188.74 g/mol
Storage Temperature:Ambient