Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethoxyphenylboronic acid
3,5-Dimethoxyphenylboronic acid ≥98%
3,5-Dimethoxyphenylboronic acid ≥98%
:
Thermo Fisher Scientific
3,5-Dimethoxyphenylboronic acid ≥98%
:
Thermo Fisher Scientific
:
192182-54-0
:
:
C₈H₁₁BO₄
:
MFCD03095127
:
205…207 °C
:
181.98 g/mol
:
GHS07
:
Ambient
msds_ALFAH52965
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years