2,5-Dibromotoluene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-59-8
:
Formula:C₇H₆Br₂
MDL Number:MFCD00000090
:249.93 g/mol
Boiling Pt:135…136 °C (35 mmHg)
Melting Pt:5…6 °C
Density:1.81 g/cm³
Flash Pt:>110 °C
EINECS:210-437-0
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6000 to 1.6040 (20°C, 589 nm)
- GC:≥97.5 %