
1,2-Dibromo-1-phenylethane
Catalog # MOLEM85625115
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:263.96 g/mol
Boiling Pt:139…141 °C (15 mmHg)
Melting Pt:75…76 °C
Density:1.74 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-253-4
UN:3261
ADR:8,III
:GHS05
CAS Number:93-52-7
Formula:C₆H₅CHBrCH₂Br
MDL Number:MFCD00000138