1,1-Diphenylethylene ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 530-48-3 Molecular Formula: C14H12 Formula Weight: 180.25 Flash Point: 136°(276°F) Refractive Index: 1.6065 Melting Point: 5-7° Boiling Point: 270-271° Beilstein Reference: 1099062 Merck Reference: 14,3323 MDL No.: MFCD00008583
Synonyms: asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
- CAS Number:530-48-3
- Formula:(C₆H₅)₂CCH₂
- MDL Number:MFCD00008583
- Molecular Weight:180.25 g/mol
- Merck Index:14,3323
- Boiling Pt:270…271 °C
- Melting Pt:5…7 °C
- Density:1.023
- Flash Pt:136 °C (276 °F)
- Storage Temperature:Ambient
- EINECS:208-482-6
4 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 530-48-3 Molecular Formula: C14H12 Formula Weight: 180.25 Flash Point: 136°(276°F) Refractive Index: 1.6065 Melting Point: 5-7° Boiling Point: 270-271° Beilstein Reference: 1099062 Merck Reference: 14,3323 MDL No.: MFCD00008583
Synonyms: asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene
- CAS Number:530-48-3
- Formula:(C₆H₅)₂CCH₂
- MDL Number:MFCD00008583
- Molecular Weight:180.25 g/mol
- Merck Index:14,3323
- Boiling Pt:270…271 °C
- Melting Pt:5…7 °C
- Density:1.023
- Flash Pt:136 °C (276 °F)
- Storage Temperature:Ambient
- EINECS:208-482-6
Documents
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



