Order Entry
Export
ContactUsLinkComponent
Organic reference standard, Benzene 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Benzene 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA10535000ME
 :  
Organic reference standard, Benzene 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA10535000ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    71-43-2
  • Description:
    Benzene 100 µg/ml in methanol
  • Formula:
    C₆H₆
  • Description:
    Benzene