4,5-Difluoroanthranilic acid, Sigma-Aldrich®
Catalog # SIAL399760-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:173.12 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:83506-93-8
Formula:C₇H₅F₂NO₂
MDL Number:MFCD00077509