Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Hydroxybiphenyl-4-carboxylic acid
4'-Hydroxybiphenyl-4-carboxylic acid ≥99%
4'-Hydroxybiphenyl-4-carboxylic acid ≥99%
:
Thermo Fisher Scientific
4'-Hydroxybiphenyl-4-carboxylic acid ≥99%
:
Thermo Fisher Scientific
:
58574-03-1
:
:
C₁₃H₁₀O₃
:
MFCD00059078
:
214.22 g/mol
:
292…295 °C
:
GHS07
:
Ambient
msds_ALFAB21946
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years