Order Entry
Export
Orders LinkContactUsLinkComponent
α-Ionone

α-Ionone

Glass bottle, 25 mL
Catalog # ROTH3950.1
Supplier:   Roth Carl
α-Ionone
Catalog # ROTH3950.1
Supplier:  Roth Carl
CAS Number:  127-41-3
α-Ionone

Glass bottle, 25 mL

CAS Number:  127-41-3
Price
40,40 €
Log in for availability
1 * 25 mL
Product Details & Documents
Synonyms:  (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one
  • CAS Number:127-41-3
  • Formula:C₁₃H₂₀O
  • MDL Number:MFCD00001565
  • Molecular Weight:192.3 g/mol
  • EINECS:204-841-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL

Specifications