α-Ionone
Glass bottle for solids, 100 g
MOLEM87966374
: Molekula
Jump to:
Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one
- CAS Number:127-41-3
- Formula:C₁₃H₂₀O
- MDL Number:MFCD00001565
- Molecular Weight:192.3 g/mol
- EINECS:204-841-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



