(E)-β-Ionone
Glass bottle for solids, 1 g
Catalog # ROTH3713.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone
- CAS Number:79-77-6
- Formula:C₁₃H₂₀O
- MDL Number:MFCD00001549
- Molecular Weight:192.30 g/mol
- EINECS:201-224-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



