Order Entry
Export
Orders LinkContactUsLinkComponent
Serotonin hydrochloride

Serotonin hydrochloride

Glass bottle for solids, 1 g
Catalog # MOLE29525281-1G
Supplier:   Molekula
Serotonin hydrochloride
Catalog # MOLE29525281-1G
Supplier:  Molekula
CAS Number:  153-98-0
Serotonin hydrochloride

Glass bottle for solids, 1 g

CAS Number:  153-98-0
Price
73,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  5-Hydroxytryptamine Hydrochloride
  • CAS Number:153-98-0
  • Formula:C₁₀H₁₃ClN₂O
  • MDL Number:MFCD00012686
  • Molecular Weight:212.68 g/mol
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications