6-Methoxyoxindole
Glass bottle for solids, 5 g
Catalog # COBBIN-0085-5G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:7699-19-6
- Formula:C₉H₉NO₂
- MDL Number:MFCD00239414
- Molecular Weight:163.18 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



