1-Boc-piperazine
Glass bottle for solids, 5 g
Catalog # MOLE36721814-5G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate
- CAS Number:57260-71-6
- Formula:C₉H₁₈N₂O₂
- MDL Number:MFCD00075265
- Molecular Weight:186.25 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



