Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dicyclopropyl ketone

Glass bottle for solids, 100 g
Catalog # MOLE13129545-100G
Supplier:   Molekula
Dicyclopropyl ketone
Catalog # MOLE13129545-100G
Supplier:  Molekula
CAS Number:  1121-37-5
Dicyclopropyl ketone

Glass bottle for solids, 100 g

CAS Number:  1121-37-5
Price
355,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Dicyclopropylketone, Dicyclopropylmethanone
  • CAS Number:1121-37-5
  • Formula:C₇H₁₀O
  • MDL Number:MFCD00001276
  • Molecular Weight:110.16 g/mol
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications