Orders LinkContactUsLinkComponent
Sulphamide ≥99%
Sulphamide ≥99%
Sulphamide ≥99%
Sulphamide ≥99%
Purity: 99% CAS No.: 7803-58-9 Molecular Formula: SO2(NH2)2 Formula Weight: 96.11 Melting Point: 91-92° Solubility: Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol MDL No.: MFCD00011606
 :  Sulphuric diamide
  •  :7803-58-9
  •  :H₄N₂O₂S
  •  :MFCD00011606
  •  :96.11 g/mol
  •  :14,8922
  •  :91…92 °C
  •  :1.61
  •  :Ambient
  •  :232-262-9
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    2,5 g
    1 * 2,5 g
    1 * 2,5 g - 44,80 €
    44,80 €
    1 * 2,5 g
       44,80 €
  • Glass bottle for solids
    10 g
    1 * 10 g
    1 * 10 g - 96,50 €
    96,50 €
    1 * 10 g
       96,50 €
  • Glass bottle for solids
    50 g
    1 * 50 g
    1 * 50 g - 356,00 €
    356,00 €
    1 * 50 g
       356,00 €