Sulphamide 98+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Sulphuric diamide
- CAS Number:7803-58-9
- Formula:H₄N₂O₂S
- MDL Number:MFCD00011606
- Molecular Weight:96.11 g/mol
- Melting Pt:87…91 °C
- Density:1.61 g/cm³
- Storage Temperature:Ambient
- EINECS:232-262-9
3 Options Available Below
Product Details & Documents
Synonyms: Sulphuric diamide
- CAS Number:7803-58-9
- Formula:H₄N₂O₂S
- MDL Number:MFCD00011606
- Molecular Weight:96.11 g/mol
- Melting Pt:87…91 °C
- Density:1.61 g/cm³
- Storage Temperature:Ambient
- EINECS:232-262-9
Documents
ACRO19902
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to practically white Crystalline powder (Free from visible impurities)
- Infrared spectrum:Conforms
- Melting point:89.0 to 93.0 °C
- Loss on drying:≤0.5 % (50 °C, 2 hrs) (vacuum)
- Elemental Analysis:≥98.0 %
- Related substances:(TLC) Passes test
- pH:5.0 to 7.0 (10 % in water)



