Order Entry
Export
ContactUsLinkComponent
Erythrosin yellowish, Sigma-Aldrich®
Erythrosin yellowish, Sigma-Aldrich®
Catalog # 234079-25G
CAS Number:  
Erythrosin yellowish, Sigma-Aldrich®
Catalog # 234079-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Colour):
    Red
  • Appearance (Form):
    Conforms to Requirements
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    255 - 261 nm
  • Absorbance:
    ≥0.25
  • TLC:
    Consistent with Past Lots
  • Solubility (Turbidity):
    Clear
    10 mg/ml, H₂O
  • Solubility (Colour):
    Red

Specification Test Results

About this item

 :
 :628.07 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:251-419-2
 :GHS07
 :GHS09
Storage Temperature:Room Temperature
CAS Number:33239-19-9
Formula:C₂₀H₈I₂Na₂O₅
MDL Number:MFCD00005045