L-Threonine ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 72-19-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: 255° dec. Beilstein Reference: 1721646 Merck Reference: 14,9380 Optical Rotation: -28° (c=5 in water) MDL No.: MFCD00064270
Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Number:72-19-5
- Formula:C₄H₉NO₃
- MDL Number:MFCD00064270
- Molecular Weight:119.12 g/mol
- Merck Index:14,9380
- Melting Pt:255 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-774-1
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 72-19-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: 255° dec. Beilstein Reference: 1721646 Merck Reference: 14,9380 Optical Rotation: -28° (c=5 in water) MDL No.: MFCD00064270
Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Number:72-19-5
- Formula:C₄H₉NO₃
- MDL Number:MFCD00064270
- Molecular Weight:119.12 g/mol
- Merck Index:14,9380
- Melting Pt:255 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-774-1



