L-Threonine, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Number:72-19-5
- Formula:C₄H₉NO₃
- MDL Number:MFCD00064270
- Molecular Weight:119.12 g/mol
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:200-774-1
3 Options Available Below
Product Details & Documents
Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Number:72-19-5
- Formula:C₄H₉NO₃
- MDL Number:MFCD00064270
- Molecular Weight:119.12 g/mol
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:200-774-1



