Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,5-Dimethoxyphenol

Glass bottle for solids, 25 g
Catalog # MOLE10632563-25G
Irritant
Supplier:   Molekula
3,5-Dimethoxyphenol
Catalog # MOLE10632563-25G
Supplier:  Molekula
CAS Number:  500-99-2
3,5-Dimethoxyphenol

Glass bottle for solids, 25 g

CAS Number:  500-99-2
Price
240,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:500-99-2
  • Formula:C₈H₁₀O₃
  • MDL Number:MFCD00008388
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:294 °C (1013 hPa)
  • Melting Pt:37…39 °C
  • Storage Temperature:Ambient
  • EINECS:207-917-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications