Order Entry
Export
Orders LinkContactUsLinkComponent
L-Threonine

L-Threonine

Supplier:   Roth Carl
L-Threonine
Supplier:  Roth Carl
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
  • CAS Number:72-19-5
  • Formula:C₄H₉NO₃
  • MDL Number:MFCD00064270
  • Molecular Weight:119.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-774-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 19,80 €
    19,80 €
    1 * 25 g
       19,80 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 40,00 €
    40,00 €
    1 * 100 g
       40,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 148,00 €
    148,00 €
    1 * 500 g
       148,00 €