Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Dibromoindole
5,7-Dibromoindole, Sigma-Aldrich®
5,7-Dibromoindole, Sigma-Aldrich®
5,7-Dibromoindole, Sigma-Aldrich®
SIAL708798-1G
:
Merck
:
5,7-Dibromoindole, Sigma-Aldrich®
SIAL708798-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
274.94 g/mol
:
36132-08-8
:
C₈H₅Br₂N
:
MFCD03090642