3,4-Dimethoxyphenol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98%
CAS No.: 2033-89-8
Molecular Formula: C8H10O3
Formula Weight: 154.16
Melting Point: 79-82°
Boiling Point: 167°/14mm
MDL No.: MFCD00008390
- CAS Number:2033-89-8
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008390
- Molecular Weight:154.17 g/mol
- Boiling Pt:167 °C (14 mmHg)
- Melting Pt:79…82 °C
- Storage Temperature:Ambient
- EINECS:217-995-4
3 Options Available Below
Product Details & Documents
Purity: 98%
CAS No.: 2033-89-8
Molecular Formula: C8H10O3
Formula Weight: 154.16
Melting Point: 79-82°
Boiling Point: 167°/14mm
MDL No.: MFCD00008390
- CAS Number:2033-89-8
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008390
- Molecular Weight:154.17 g/mol
- Boiling Pt:167 °C (14 mmHg)
- Melting Pt:79…82 °C
- Storage Temperature:Ambient
- EINECS:217-995-4



