3,4-Dimethoxyphenol 97%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:2033-89-8
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008390
- Molecular Weight:154.17 g/mol
- Boiling Pt:120 - 122 °C (0,2 -rr)
- Melting Pt:77…82 °C
- Storage Temperature:Ambient
- EINECS:217-995-4
2 Options Available Below
Product Details & Documents
- CAS Number:2033-89-8
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008390
- Molecular Weight:154.17 g/mol
- Boiling Pt:120 - 122 °C (0,2 -rr)
- Melting Pt:77…82 °C
- Storage Temperature:Ambient
- EINECS:217-995-4
Documents
ACRO18670
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Off-white to light beige Crystalline powder (May darken on storage)
- Infrared spectrum:Conforms
- Melting point:77 to 82 °C
- GC:≥96.0 %



