1-Adamantaneethanol, Sigma-Aldrich®
Catalog # SIAL188115-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.29 g/mol
Boiling Pt:270 °C (1013 hPa)
Melting Pt:98.5…100 °C
:M-Clarity™ quality level = MQ100
EINECS:228-353-8
CAS Number:6240-11-5
Formula:C₁₂H₂₀O
MDL Number:MFCD00074756