Atrazine
250 mg
Catalog # MOLEM80703758
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
- CAS Number:1912-24-9
- Formula:C₈H₁₄ClN₅
- MDL Number:MFCD00041810
- Molecular Weight:215.69 g/mol
- Merck Index:12,00902
- Boiling Pt:369 °C (1013 hPa)
- Melting Pt:171…174 °C
- Density:1.2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-617-8
- ADR:9,III
- :Density: 1.2 g/cm³ (20 °C)
- UN:3077
Specifications
Specifications
- Pk:250 mg



