1,3-Dimethoxybenzene 99%
:151-10-0
:
:C₈H₁₀O₂
:MFCD00008384
:138.17 g/mol
:85…87 °C (7 mmHg)
:–52 °C
:1.05 g/cm³
:87 °C
:Ambient
:205-783-4
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5230 to 1.5250 (20°C, 589 nm)
- GC:≥98.5 %