Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diethoxybenzene 95%

1,3-Diethoxybenzene 95%

1,3-Diethoxybenzene 95%
Certificates
1,3-Diethoxybenzene 95%
1,3-Diethoxybenzene 95%

3 Options Available Below

Product Details & Documents

Purity: 95% CAS No.: 2049-73-2 Molecular Formula: C10H14O2 Formula Weight: 166.22 Refractive Index: 1.5095 Melting Point: 11-12° Boiling Point: 234-236° Beilstein Reference: 2517730 MDL No.: MFCD00015145

  • CAS Number:2049-73-2
  • Formula:C₁₀H₁₄O₂
  • MDL Number:MFCD00015145
  • Molecular Weight:166.22 g/mol
  • Boiling Pt:234…236 °C
  • Melting Pt:11…12 °C
  • Density:1.010
  • Storage Temperature:Ambient
  • EINECS:218-071-3
Product Family Options

Product Information