α,α,α-Trifluoro-o-toluidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-17-5
:
Formula:C₇H₆F₃N
MDL Number:MFCD00007718
Boiling Pt:175 °C (1013 hPa)
Melting Pt:–33 °C
Density:1.282 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-806-7
UN:2942
ADR:6.1,III
:161.13 g/mol