Order Entry
Export
Orders LinkContactUsLinkComponent
N,N-Dimethylacetamide

N,N-Dimethylacetamide

Glass bottle, 1 L
Catalog # ROTHHN45.1
Health
Irritant
Supplier:   Roth Carl
N,N-Dimethylacetamide
Catalog # ROTHHN45.1
Supplier:  Roth Carl
CAS Number:  127-19-5
N,N-Dimethylacetamide
N,N-Dimethylacetamide

Glass bottle, 1 L

CAS Number:  127-19-5
Price
338,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
  • CAS Number:127-19-5
  • Formula:CH₃CON(CH₃)₂
  • MDL Number:MFCD00008686
  • Molecular Weight:87.12 g/mol
  • Merck Index:13,03254
  • Boiling Pt:166.1 °C (1013 hPa)
  • Melting Pt:–20 °C
  • Density:0.937 g/cm³ (20 °C)
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • EINECS:204-826-4
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications