1,2-Epoxybutane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-88-7
:
Formula:C₄H₈O
MDL Number:MFCD00005153
:72.11 g/mol
Boiling Pt:63 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.83 g/cm³ (20 °C)
Flash Pt:–26 °C
Storage Temperature:Ambient
UN:3022
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:203-438-2