Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,2-difluoro-1,3-benzodioxole, Sigma-Aldrich®
Irritant
4-Bromo-2,2-difluoro-1,3-benzodioxole, Sigma-Aldrich®
Catalog # SIAL714542-1G
Supplier:  Merck
CAS Number:  
Irritant
4-Bromo-2,2-difluoro-1,3-benzodioxole, Sigma-Aldrich®
Catalog # SIAL714542-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237 g/mol
 :GHS07
CAS Number:144584-66-7
Formula:C₇H₃BrF₂O₂
MDL Number:MFCD01631385