


3,4-Dinitrophenol stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:577-71-9
:
Formula:C₆H₄N₂O₅
:184.11 g/mol
Boiling Pt:419 °C (1013 hPa)
Melting Pt:132 °C
Density:1.672 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-415-3
UN:1320
ADR:4.1,I
:Moistened with water
:GHS06
:GHS08
:GHS09