
2,4-Dinitroanisole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:119-27-7
:
Formula:C₇H₆N₂O₅
MDL Number:MFCD00035745
Boiling Pt:351 °C (1013 hPa)
Melting Pt:88 °C
Density:1.34 g/cm³ (20 °C)
:198.14 g/mol
EINECS:204-310-9
:GHS07