Order Entry
Export
Orders LinkContactUsLinkComponent
Nitrobenzene 99+% ACS

Nitrobenzene 99+% ACS

Toxic
Health
Environment
Nitrobenzene 99+% ACS
Certificates
Nitrobenzene 99+% ACS

3 Options Available Below

Product Details & Documents
  • CAS Number:98-95-3
  • Formula:C₆H₅NO₂
  • MDL Number:MFCD00007043
  • Molecular Weight:123.11 g/mol
  • Merck Index:13,06621
  • Boiling Pt:210.8 °C (1013 hPa)
  • Melting Pt:5…6 °C
  • Density:1.205 g/cm³
  • Flash Pt:88 °C
  • Storage Temperature:Ambient
  • EINECS:202-716-0
  • UN:1662
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5500 to 1.5540 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.005 %
  • Chloride (Cl):
    ≤5 ppm
  • Water soluble titr. acid:
    ≤0.0005 meq/g
Product Family Options

Product Information