Order Entry
Export
ContactUsLinkComponent
1,4-Dinitrobenzene
Toxic
Health
Environment
1,4-Dinitrobenzene
Catalog # EHERC12783200
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
Environment
1,4-Dinitrobenzene
Catalog # EHERC12783200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :168.11 g/mol
Merck Index:13,03301
Boiling Pt:299 °C (1013 hPa)
Melting Pt:172 °C
Density:1.625 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-833-7
UN:3443
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:100-25-4
Formula:C₆H₄N₂O₄
MDL Number:MFCD00007314