


1,4-Dinitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-25-4
:
Formula:C₆H₄N₂O₄
MDL Number:MFCD00007314
:168.11 g/mol
Merck Index:14,3273
Boiling Pt:299 °C
Melting Pt:172…176 °C
Density:1.625
Flash Pt:150 °C (302 °F)
Storage Temperature:Ambient
EINECS:202-833-7
UN:3443
ADR:6.1,II
:GHS06
:GHS08
:GHS09