


1,4-Dinitrobenzene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-25-4
:
Formula:C₆H₄N₂O₄
MDL Number:MFCD00007314
:168.11 g/mol
Merck Index:13,03301
Boiling Pt:299 °C (1013 hPa)
Melting Pt:173…175 °C
Density:1.63 g/cm³
Flash Pt:150 °C
Storage Temperature:Ambient
EINECS:202-833-7
UN:3443
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Ochre to orange Powder or crystals
- Infrared spectrum:Conforms
- Melting point:171 to 175 °C
- GC:≥97.5 %