Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethoxyaniline

3,5-Dimethoxyaniline

Glass bottle for solids, 25 g
Catalog # MOLE37359094-25G
Irritant
Supplier:   Molekula
3,5-Dimethoxyaniline
Catalog # MOLE37359094-25G
Supplier:  Molekula
CAS Number:  10272-07-8
3,5-Dimethoxyaniline

Glass bottle for solids, 25 g

CAS Number:  10272-07-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:10272-07-8
  • Formula:C₈H₁₁NO₂
  • MDL Number:MFCD00008392
  • Molecular Weight:153.18 g/mol
  • Boiling Pt:303 °C (1013 hPa)
  • Melting Pt:66…68 °C
  • Storage Temperature:Ambient
  • EINECS:233-616-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications