
3,4-Dimethoxyaniline
Supplier: CHEMPUR
Certificates
About this item
CAS Number:6315-89-5
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00008394
Boiling Pt:296 °C (1013 hPa)
Melting Pt:85…88 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:228-647-6
UN:2811
ADR:6.1,III
:153.18 g/mol
:GHS07