

3,4-Dimethoxyaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6315-89-5
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00008394
Boiling Pt:174…176 °C (22 mmHg)
Melting Pt:86…89 °C
Flash Pt:>110 °C (230 °F)
EINECS:228-647-6
UN:2811
ADR:6.1,III
:153.18 g/mol
:GHS06
:GHS08
Storage Temperature:Ambient