2,5-Dimethoxyaniline
Catalog # MOLE72723280-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:153.18 g/mol
Boiling Pt:270 °C (1013 hPa)
Melting Pt:81 °C
Storage Temperature:Ambient
EINECS:203-040-9
UN:2811
ADR:6.1,III
CAS Number:102-56-7
Formula:C₈H₁₁NO₂
MDL Number:MFCD00008368