


2,6-Dinitroaniline
Catalog # MOLE29109024-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:183.12 g/mol
Merck Index:13,03299
Melting Pt:136 °C
Density:1.629 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-108-1
UN:1596
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:606-22-4
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007148