
N,N-Dibutylaniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:613-29-6
:
Formula:(C₆H₅)N(CH₂CH₂CH₂CH₃)₂
MDL Number:MFCD00035799
:205.34 g/mol
Boiling Pt:269…270 °C
Melting Pt:–32 °C
Density:0.906
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:210-335-6
UN:2810
ADR:6.1,III
:GHS06