1,4-Diiodobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:624-38-4
:
Formula:C₆H₄I₂
MDL Number:MFCD00001054
:329.91 g/mol
Boiling Pt:285 °C
Melting Pt:128…132 °C
Density:2.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-842-2
:GHS07
Specifications
- Environmentally Preferable: