Order Entry
Export
ContactUsLinkComponent
1,4-Diiodobenzene 98%
Irritant
1,4-Diiodobenzene 98%
Certificates
undefined
Irritant
1,4-Diiodobenzene 98%

Specification Test Results

  • Appearance:
    White to beige to tan Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    128 to 133 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information