1,3-Diiodobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-00-6
:
Formula:C₆H₄I₂
MDL Number:MFCD00041731
:329.91 g/mol
Boiling Pt:285 °C
Melting Pt:35…37 °C
Density:2.470
Flash Pt:>110 °C (230 °F)
EINECS:210-921-1
:GHS07
Storage Temperature:Ambient