1,3-Diiodobenzene 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:626-00-6
- Formula:C₆H₄I₂
- MDL Number:MFCD00041731
- Molecular Weight:329.91 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:33…37 °C
- Density:2.47 g/cm³
- Storage Temperature:Refrigerator
- EINECS:210-921-1
2 Options Available Below
Product Details & Documents
- CAS Number:626-00-6
- Formula:C₆H₄I₂
- MDL Number:MFCD00041731
- Molecular Weight:329.91 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:33…37 °C
- Density:2.47 g/cm³
- Storage Temperature:Refrigerator
- EINECS:210-921-1
Documents
ACRO40774
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to beige Powder or crystals or low melting crystalline mass
- Infrared spectrum:Conforms
- Melting point:33 to 37 °C
- GC:≥97.5 %



